Learn More
VACUUBRAND™ DCP 3000 Vacuum Gauge
The DCP 3000 is an outstanding vacuum gauge for rough and fine vacuum
Brand:  VACUUBRAND™ 20636155
Additional Details : Weight : 0.01000kg
Product Code. 11901002
  • 6600.00 DKK / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
The DCP 3000 is an outstanding vacuum gauge for rough and fine vacuum. A large illuminated display shows the measured values of all sensors by a simple turn of a button. The VSK 3000 vacuum sensor with ceramic diaphragm measures from atmospheric pressure down to 0.1 mbar. It displays absolute pressure independent of gas type, is very corrosion resistant and has an outstanding accuracy and long term stability. Simultaneous connection of up to eight vacuum sensors allows flexible measuring on the spot. The DCP 3000 communicates with external components via the VACUU·BUS™ digital bus system. It is characterized by fully automatic configuration, unified plug connections and cable lengths of up to 30 m. In addition the DCP 3000 offers relative pressure measurements (VSK 3000 as reference) and a data logger feature for up to 32,000 data values.
Specifications
Specifications
Vacuum Gauge