Learn More
Invitrogen™ Custom Primer, Plate
Custom Primer, Plate
Brand: Invitrogen™ 10629178
3.73 DKK valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.00500kg
Description
Order multiple custom DNA oligos in plate format by selecting either 96 or 384-wells formats. Simply download either the 96-well Excel template or 384-well Excel template, enter your sequence and plate location information, and upload in the appropriate format.Specifications
Normal or Reversed-phase Chromatography, HPLC or PAGE | |
Custom Plate Primer | |
HPLC |
Custom Plate Primer, 200 nmole, Purified by Normal or Reversed-phase Chromatography, HPLC or PAGE, Used to Design a Custom DNA Oligonucleotide, With any of a Variety of 3' and 5' Modification | |
200 nmol | |
Design a Custom DNA Oligonucleotide, with any of a variety of 3' and 5' Modification |