Learn More
Multiroir Controlec™ Crystal Polystyrene Storage Drawers
View the contents of Multiroir Controlec™ Crystal Polystyrene Storage Drawers clearly due to their construction of crystal clear polystyrene. The drawers also feature a slide assembly, with dividers available to order separately.
190.00 DKK - 365.00 DKK
Specifications
Description | Handle type: Slider assembly |
---|---|
Color | Transparent |
Material | Polystyrene |
Shape | Rectangular |
Product Type | Drawer |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11768303
|
Multiroir Controlec™
11103MU07000C |
7 kg |
365.00 DKK
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11798283
|
Multiroir Controlec™
11101MUO19TR |
1.9 kg |
190.00 DKK
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Total visibility
- Crystal polystyrene transparent - like glass
- Slide assembly
- Drawer separators available separately
Specifications
Handle type: Slider assembly | |
Polystyrene | |
Drawer |
Transparent | |
Rectangular | |
1 Each |