Learn More
Corning™ TC-Treated Culture Dishes
Tissue-culture (TC) treated for optimum attachment and growth of anchorage-dependent cells. Corning™ TC-Treated Culture Dishes provide an ideal solution for growing and maintaining cells in vitro.
1729.48 DKK - 3498.07 DKK
Specifications
Closure Color | Clear |
---|---|
Closure Included | Yes |
Closure Material | Polystyrene |
Bottom Shape | Flat |
Color | Clear |
Product Code | Brand | Shape | Culture Area | Description | Gridded | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Shape | Culture Area | Description | Gridded | Price | Quantity & Availability | ||
10310272
|
Corning™
430196 |
Round | 21 cm2 | 51.4 dia. x 15 mm H | Gridded |
2598.98 DKK
Pack of 500 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||
10350602
|
Corning™
431110 |
Square | 500 cm2 | 245 L x 245 W x 25 mm H | Not Gridded |
1797.89 DKK
Pack of 16 |
In Stock
Log in to see stock. |
||
10095381
|
Corning™
430293 |
Round | 55 cm2 | 80.5 dia. x 20 mm H | Not Gridded |
3414.54 DKK
Pack of 480 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||
10556661
|
Corning™
430165 |
Round | 9 cm2 | 34.4 dia. x 10 mm H | Not Gridded |
2136.95 DKK
Pack of 500 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||
10634141
|
Corning™
430166 |
Round | 21 cm2 | 52.1 dia. x 15 mm H | Not Gridded |
2336.07 DKK
Pack of 500 |
In Stock
Log in to see stock. |
||
10075371
|
Corning™
430167 |
Round | 55 cm2 | 83.8 dia. x 20 mm H | Not Gridded |
3498.07 DKK
Pack of 500 |
In Stock
Log in to see stock. |
||
10117320
|
Corning™
430599 |
Round | 148 cm2 | 134.5 dia. x 25 mm H | Not Gridded |
1729.48 DKK
Pack of 60 |
In Stock
Log in to see stock. |
||
Description
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment
- Ultra-Low Attachment surface dishes feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation
- 10 dishes/bag are available for 100 mm dishes (Cat. No. 430293)
- 245 mm square dishes offer 500 cm2 growth surface
- Stacking beads aid in handling
- Vents provide consistent gas exchange
- Manufactured from optically clear virgin polystyrene
- Sterile
- Nonpyrogenic
Dish style (mm) = actual growth surface diameters: 35 mm dish = 34.4 mm; 60 mm dish = 52.1 mm; 100 mm dish = 83.8 mm; 150 mm dish = 139.1 mm.
Cat. no. 431110, Square dishes with interior bottom dimensions of 224 x 224 mm.
Order Info
Package quantities differ. Check ordering table for details.
Specifications
Clear | |
Polystyrene | |
Clear | |
Polystyrene | |
Cell Culture | |
Lid |
Yes | |
Flat | |
Single-use | |
Sterile | |
Cell Culture Petri Dish |