Learn More
3M™ Comply Disposable Bowie & Autoclave Test Pack
Fast, efficient and easy to use. Paper can be re-used for note-taking
Brand: 3M™ 7000066392
Additional Details : Weight : 1.00000kg
Description
With unique color chemistry, the 3M™ Comply™ disposable Bowie and Dick type autoclave test pack conforms to BS EN 867-4 and carries the BSI KiteMark™.- Fast, efficient and disposable
- Simple and easy to read
- Stronger colors and higher definition
- Accurate, consistent, reliable and reproducible
- Easy record storage
- Saves time
- Paper can be re-used for note-taking For single use in porous load sterilisers for daily Bowie and Dick tests.
Specifications
Autoclavable | |
BS EN 867-4 | |
Disposable Bowie and Autoclave Test | |
20 Pk. |