Learn More
Macherey-Nagel™ Chromafix™ Dry (Na2SO4) Cartridge
935.00 DKK - 1470.00 DKK
Specifications
Base Material | Dry (Na2 SO4) |
---|---|
For Use With (Application) | Removal of Traces of Water From Organic Solutions |
Type | Chromafix Dry (Na2 SO4) Cartridge |
Quantity | 50 Pack |
Product Code | Brand | Bed Weight | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight | Price | Quantity & Availability | |||||
10639771
|
Macherey-Nagel™
731853 |
1500 mg |
1185.00 DKK
Pack of 50 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
10639961
|
Macherey-Nagel™
731854 |
2800 mg |
1470.00 DKK
Pack of 50 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
10441282
|
Macherey-Nagel™
731852 |
780 mg |
935.00 DKK
Pack of 50 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
Specifications
Dry (Na2 SO4) | |
Removal of Traces of Water From Organic Solutions | |
Chromafix Dry (Na2 SO4) Cartridge | |
50 Pack |