Learn More
Macherey-Nagel™ Chromabond™ Leur Fitting
790.00 DKK
Specifications
For Use With (Equipment) | Lid, Vacuum Manifolds |
---|---|
Type | Luer Fitting |
Quantity | 12 Pack |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10647548
|
Macherey-Nagel™
730184.12 |
Male |
790.00 DKK
Pack of 12 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
10502499
|
Macherey-Nagel™
730183.12 |
Female |
790.00 DKK
Pack of 12 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Specifications
Lid, Vacuum Manifolds | |
Luer Fitting | |
12 Pack |