Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Calibrated Density Bottles
Calibrated density bottle made with Borosilicate glass that is ideal for use in most research, scientific, and other types of laboratory environments.
434.00 DKK - 560.00 DKK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10663561
|
BRAND™
43308 |
10 mL |
448.00 DKK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
10693371
|
BRAND™
43305 |
5 mL |
434.00 DKK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
10748171
|
BRAND™
43328 |
50 mL |
560.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
10747981
|
BRAND™
43320 |
25 mL |
490.00 DKK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
Description
- Calibrated density bottle made with Borosilicate glass
- DIN ISO 3507, type Gay-Lussac
- Calibrated to contain (TC, In)
- Includes individual certificate
- Stopper NS 10/19 with capillary
- Top of stopper ground and polished
- The volume in cm³ is specified to a precision of 3 decimal places
Specifications
Borosilicate Glass |