missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK16, Human, Matched Antibody Pair (AP11), Abnova™
Brand: Abnova H00008576-AP11.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-STK16 (100μg); Detection antibody: mouse monoclonal anti-STK16, IgG1 Kappa (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human STK16.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Specifications
STK16 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human STK16. | |
FLJ39635/KRCT/MPSK/PKL12/TSF1 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-STK16 (100 μg) 2. Detection antibody: mouse monoclonal anti-STK16, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
STK16 | |
STK16 | |
Affinity Purified | |
RUO | |
8576 | |
Human |