Learn More
anti-IL32, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00009235-AP22.1Set
Product Code. 16132984
  • 6285.00 DKK / 1 set
Explore more special offers
Includes: Capture antibody: rabbit MaxPab affinity purified polyclonal anti-IL32 (100μg) , Detection antibody: mouse purified polyclonal anti-IL32 (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
IL32
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IL32.
IL-32alpha/IL-32beta/IL-32delta/IL-32gamma/NK4/TAIF/TAIFa/TAIFb/TAIFc/TAIFd
Mouse, Rabbit
RUO
9235
Human
Liquid
ELISA
Unconjugated
IL32
IL32
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-IL32 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-IL32 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Special Offers
For Research Use Only