Learn More
Heathrow Scientific™ Aluminum Test Tube Basket
Wash and store test tubes. Heathrow Scientific™ Aluminum Test Tube Basket can withstand repeated exposure to autoclaves.
224.00 DKK - 269.00 DKK
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Aluminum |
For Use With (Application) | Washing, Autoclaving, Storing Test Tubes |
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
12327569
|
Heathrow Scientific™
HS20341B |
256 x 152 x 152mm |
269.00 DKK
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
12317569
|
Heathrow Scientific™
HS20341A |
152 x 152 x 152mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12337569
|
Heathrow Scientific™
HS20341C |
332 x 228 x 178mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Constructed of lightweight aluminum
- Ideal for washing and storing test tubes
- Withstands repeated autoclaving and use in the lab
Specifications
Autoclavable | |
Aluminum | |
Washing, Autoclaving, Storing Test Tubes |