Learn More
Masterflex™ 4-way Stopcock
Use these stopcocks for a variety of applications. Cole-Parmer™ 4-way Stopcocks feature a PC body and an HDPE stem.
307.00 DKK - 336.00 DKK
Specifications
Type | Stopcock |
---|---|
Material | Polycarbonate, HDPE |
Plug Material | Polycarbonate and HDPE |
Outlet Port | Male Luer Lock or A Male Luer Slip |
Product Code | Brand | Connectors | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connectors | Price | Quantity & Availability | |||||
11742683
|
Masterflex™
30600-04 |
Male Luer Lock |
336.00 DKK
Pack of 10 |
N/A | |||||
Currently Unavailable: We have no information when this product will be available.
View alternative products
|
11762673
|
Masterflex™
30600-03 |
Male Luer Slip |
307.00 DKK
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
Description
Use these stopcocks for a variety of applications. Cole-Parmer™ 4-way Stopcocks feature a PC body and an HDPE stem.
- Four-way stopcock
- Choose stopcocks with either a male luer lock or a male luer slip
- Handle rotates 360 degrees on four-way stopcocks
Specifications
Stopcock | |
Polycarbonate, HDPE | |
Polycarbonate and HDPE | |
Male Luer Lock or A Male Luer Slip |