Learn More
3M™ Qualitative Fit-Test Kit Accessories
For use with 3M™ Qualitative Fit Test Apparatus
398.31 DKK - 1532.65 DKK
Product Code | Brand | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||
15749767
|
3M™
7000051788 |
Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter | Respirator Test Hood |
398.31 DKK
Pack of 2 Qty Price Break Available |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
15135157
|
3M™
7000001945 |
Quantitative Fit Testing | Fit Test Adapter |
1510.00 DKK
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
12973731
|
3M™
7000001897 |
Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter | Nebulizer |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12993731
|
3M™
7000051789 |
Provide secure hood environment | Collar |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Test kits and accessories designed to help your organization meet OSHA fit testing requirements for respirators
- The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
- 3M Catalog# 601, FT-13, FT-14, FT-15