Learn More
DWK Life Sciences DURAN™ Polyethylene Stopper
The ergonomically shaped stoppers are suitable for closing volumetric flasks, mixing cylinders and standing bottles.
Brand:  DWK Life Sciences 292050604
89.11 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.06000kg
Product Code. 15562245
  • 93.80 DKK / Pack of 10
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Description
Description
The stoppers have an ergonomic grip that especially makes the opening and closing of volumetric flasks, mixing cylinders and reagent bottles easier and more secure. For optimal flask sealing the tapered bodies of the stoppers have moulded ridges. Coloured inserts in the different sizes of stoppers allow the easy identification of the corresponding ground (ns/TS) neck size. The colour identification system used for the inserts is identical to that used for the KECKTM interchangeable glassware clips.
  • Supplied with ergonomic polyethylene stopper for easy and safe tightening or removal
  • Taper with several grooves ensures optimal flask sealing
  • Coloured inserts in the different sizes of stoppers allow the easy identification of the corresponding standard ground neck size
  • The colour identification system used for the inserts is identical to that used for the KECK™ interchangeable glassware clips
  • The batch indicator represents the production period (year and month)
  • With material designation for polyethylene PE04 on the underside of the product
Specifications
Specifications
Yellow
10/Pk.
No
Preparation and storage of standard solutions; color coding and identification; Avoidance of solvent evaporation
Polyethylene